Would you like to log in again?
VWR® Aerosol Barrier Pipet Tips, Standard Line
Supplier: VWR International
10017-104 10017-072 10017-100 10017-074 10017-102
10017-104CS 670.57 USD InStock
10017-104 10017-072 10017-100 10017-074 10017-102
VWR® Aerosol Barrier Pipet Tips, Standard Line
Pipet Tips
VWR® Standard Line Pipet Tips utilize the industry standard in aerosol barrier technology.
  • Translucent packaging for easy vizualization of tips
  • Built with stability for ease of use when multi-channel pipetting
  • Back hinge makes it easy to open and close with one hand
  • Certified free of contaminants for the most sensitive laboratory protocols
These tips are conveniently racked in the most widely used style of rack on the market.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty