    Sort by:
  • Search Within Results
Plumb Bobs
https://us.vwr.com/stibo/smallweb/std.lang.all/96/59/18989659.jpg
A plumb bob is a weight that is suspended from a string and used as a vertical line of reference, used to ensure that constructions are plumb, or vertical. They are used in a variety of instruments to set the instrument exactly over a fixed survey marker, or to transcribe positions onto the ground for placing a marker. Another popular use for these tools is to determine the center of gravity of an irregular shape.
SearchPresentationType-VERTICAL
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (470123-240)
Supplier:  Wards
Description:   PLUMB BOB HEXAGONAL - 8OZ.