Would you like to log in again?
Module 3: DNA Analysis & Cloning
Supplier: Edvotek
403
470136-544KT 3323.29 USD InStock
470136-544
Module 3: DNA Analysis & Cloning
Educational Materials Biology Educational Materials Biology Textbooks
Features a variety of stimulating experiments to help students learn about the science of DNA and its application. The power of molecular cloning and restriction enzyme is remarkable.
Curriculum Module Includes:
• Micropipetting Basics
• Principles and Practice of Agarose Gel Electrophoresis (2)
• Restriction Enzyme Cleavage of Plasmid and Lambda DNA
• Size Determination of DNA Restriction Fragments (2)
• Mapping of Restriction Sites on Plasmid DNA (2)
• Mini-Prep Isolation of Plasmid DNA
• Isolation of E.coli Chromosomal DNA
• Cleavage of DNA With Restriction Enzymes (2)
• Transformation of E. coli with GFP and BFP
• Blue/White Cloning of a DNA Fragment and Assay of B-galactosidase (2)
• Construction and Cloning of a DNA Recombinant (2)
• Purification of the Restriction Enzyme Eco RI (2)
• Cloning of a PCR Amplified Gene (2)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty