Would you like to log in again?
Exploring Aquifers and Water Flow Lab Activity
Supplier: VWR
WARD470023-726 WARD470023-724
470023-726KT 199.99 USD InStock
470023-726 470023-724
Exploring Aquifers and Water Flow Lab Activity
Educational Classroom Kits and Activities
Study Key Groundwater Principles
  • Demonstrates how water flows through an aquifer
  • Determine how elevation effects water flow
Students construct a simple working model to demonstrate Darcy's Law and examine groundwater flow in an artesian system.
You will receive all materials, a teacher's guide, and student copymasters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty