Anti-KLRK1 Rat Monoclonal Antibody [clone: CX5]

70-5882-U100
10002-338EA 105.07 USD
10002-338
Anti-KLRK1 Rat Monoclonal Antibody [clone: CX5]
The CX5 antibody is specific for mouse NKG2D (CD314, KLRK1), a lectin-like type II transmembrane receptor protein. NKG2D functions as an activating receptor on NK cells and a co-stimulatory receptor on cytotoxic T cells, in both cases associating with the DAP10 adaptor protein. It is not expressed on resting T cells or unstimulated macrophages. NKG2D binds to its ligands, often expressed on atypical cells, including MULT1, H60 and five RAE1 variants. The CX5 antibody is reported to block binding of NKG2D to its ligands. Please choose the appropriate format for each application.

Type: Primary
Antigen: KLRK1
Clonality: Monoclonal
Clone: CX5
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR