Would you like to log in again?
Sklar-Cide28 High Level Disinfecting Solution, Sklar
Supplier: Sklar
10-1623
89022-228CS 210.56 USD InStock
89022-228
Sklar-Cide28 High Level Disinfecting Solution, Sklar
Cleaning Agents Disinfectants
Sklar-Cide28 High Level Disinfecting Solution provides an efficient and safe means for the high-level disinfection or sterilization of medical, ophthalmic, dental, and surgical devices including endoscopes and lensed instruments
As with all liquid sterilants, this product should be used for instruments and devices that are heat labile or otherwise incompatible with other methods of biologically monitored sterilization.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty