Would you like to log in again?
Disinfectant, Sklar
Supplier: Sklar
10-1644 10-1642
89022-252EA 227.86 USD InStock
89022-252 10185-050
Disinfectant, Sklar
Cleaning Agents Disinfectants
Sklar Disinfectant is a ready to use, highly effective cleaner and disinfectant for use in intensive care units and high-risk patient care areas, surgical suites, infant care units, plasmapheresis facilities and other areas where strict environmental control of cross contamination is critical
May be applied directly to surfaces or used as an immersion solution.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty