Would you like to log in again?
Compact Pin Moisture Meter, Extech
Supplier: Extech
MO50
10017-262EA 57.38 USD InStock
10017-262
Compact Pin Moisture Meter, Extech
Data Loggers Data Loggers for Education
The Compact Pin Moisture Meter displays moisture level in wood and building materials such as wall board, sheet rock, cardboard, plaster, concrete, and mortar
The Compact Pin Moisture Meter offers a wood moisture range of 5 to 50%, and a non-wood range of 1.5 to 33%.
Icons display low, medium and high levels of moisture content, with an audible alert that beeps faster as the moisture level increases. Easy-to-use, compact-sized design also features a built-in battery level check and measurement verification test.
Certifications: CE compliant.
Ordering information: Supplied complete with pins (4), protective cap, and 9V battery.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty