Would you like to log in again?
EzSmart™ Hygro-Thermometer, Extech
Supplier: Extech
RHT3
10017-260EA 80.77 USD InStock
10017-260
EzSmart™ Hygro-Thermometer, Extech
Data Loggers Temperature and Humidity Loggers
This compact Hygro-Thermometer for iOS® and Android™ smart devices measures humidity and temperature simultaneously, and calculates wet bulb and dew point temperature
A continuous graph plots humidity and temperature changes over time, with five user-settable high/low alarms for both temperature and humidity.
Certifications: CE compliant.
Ordering information: Supplied complete with built-in sensor and protective connector cap with strap.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty