Would you like to log in again?
USB Temperature Datalogger, Extech
Supplier: Extech
THD5
10017-258EA 401.74 USD InStock
10017-258
USB Temperature Datalogger, Extech
Data Loggers Temperature Loggers
Convenient credit card-sized housing with USB 2.0 connector and NTC thermistor for one-time use in datalogging accurate temperature measurements
User programmable settings include language, sample rate, start delay time, alarm delay time, high/low alarm range, and security feature. Status indication is provided via red and green LEDs. The start button begins continuous datalogging, which continues until memory is full at 8192 readings, or when stop button is pressed. Automatically generates a PDF report with data and trending graph when plugged into a PC after datalogging is completed.
Certifications: CE Compliant
Ordering information: Supplied complete with 10 datalogging units (each with a CR2032 button battery and plastic bag).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty