Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

True RMS 200A AC Open Jaw Clamp Meter, Extech

Supplier: Extech
MA260
10017-250EA 179.51 USD InStock
10017-250
True RMS 200A AC Open Jaw Clamp Meter, Extech
Data Loggers Voltage/Current Loggers
Smart Auto Sensing technology enables the meter to recognize the input and automatically switch to the correct mode of operation

For AC Voltage, Auto Sense feature evaluates input signal and adjusts input impedance to eliminate effects of ghost voltages. The unit provides True RMS for accurate readings of noisy, distorted, or non-sinusoidal waveforms. Standard features include 16mm (0.6") open jaw size, built-in non-contact voltage detector with LED indicator, 10,000 count backlit LCD display, and Smart Data Hold (freezes the reading on the display and automatically changes if a new reading is larger than 50 counts).

Certifications: UL listed. CE compliant.

Ordering information: Meter is supplied complete with test leads and two AAA batteries.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty