3000 A True RMS AC Flex Clamp Meter, Extech
Supplier: Extech
CA3018 MA3010 MA3018 CA3010
10017-238EA 342.52 USD
10017-238 10017-244 10017-242 10017-240
3000 A True RMS AC Flex Clamp Meter, Extech
Data Loggers
The 3000A True RMS AC Flex Clamp Meter measures AC current up to 3000 A. The 3000A AC Flex clamp-on adaptors are compatible with most multimeters and clamp meters with standard sized banana plug inputs.
  • Accurate readings of noisy, distorted, or non-sinusoidal waveforms; measures AC current in three ranges
  • Flexible 10" or 18" clamp jaws easily wrap around bus bars and cable bundles.
  • 7.5 mm cable diameter fits into tight spaces and around large conductors
  • With a 3000 count backlit LCD display and over-range and low battery indicator.
Green and yellow housing.
Certifications: UL listed. CE compliant.
Ordering information: Both the 10" and 18" clamp meters are supplied complete with two AAA batteries. Additionally, both the 10" and 18" clamp-on adapters are supplied complete with built-in flex clamp cable, 2m (6.6') lead with banana plugs, and two AAA batteries.