Sprague-Rappaport Diagnostic Stethoscope, Sklar
06-2304
10185-602EA 46.27 USD
10185-602
Sprague-Rappaport Diagnostic Stethoscope, Sklar
The Sprague-Rappaport Diagnostic Stethoscope features thick, dual tubing to pick up very faint sounds
This five-in-one stethoscope can be used on adult, pediatric, and infant patients. It is supplied with two diaphragms, three bells, and two additional ear pieces. The large diaphragm and the large bell are used with adult patients; a small diaphragm and small bell are used with infant and pediatric patients; and a medium bell can be used to listen for sounds in narrow or small places. This stethoscope has 55.8cm (22") black PVC tubing and an overall length of 76.2cm (30").