Would you like to log in again?
Newborn Stethoscope, Sklar
Supplier: Sklar
06-1642
10194-512EA 35.1 USD InStock
10194-512
Newborn Stethoscope, Sklar
Stethoscopes
The Newborn Stethoscope features a traditional bell/diaphragm design and is sized and acoustically tailored for newborn infants
Both the bell and the diaphragm on this instrument measure 3cm ( 1³⁄₁₆"). The overall length is 76.2cm (30") and the PVC tubing is gray.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty