Would you like to log in again?
Dual Head Cardiology Stethoscope, Sklar
Supplier: Sklar
06-1672 06-1600 06-1673 06-1671 06-1670
10185-616EA 133.62 USD InStock
10185-616 10194-496 10185-618 10185-614 10185-612
Dual Head Cardiology Stethoscope, Sklar
Stethoscopes
The Dual Head Cardiology Stethoscope allows the user to hear high, medium, and low frequency sounds
Specifically those identified with the heart, such as blood flow, clicks, and murmurs. The cardiology stethoscope has thicker, shorter PVC tubing than a standard stethoscope. This provides a shorter distance for sounds to travel and decreases interference from curled tubing that can occur with longer stethoscopes. The result is a significantly more accurate identification of sounds. The stethoscope features a stainless steel diaphragm and bell to further improve sound transmission. Available with a variety of tubing colors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty