Mouse Cell Array, ProSci Incorporated
Supplier: ProSci
17-701
10126-928PK 171.96 USD
10126-928
Mouse Cell Array, ProSci Incorporated
Slides Microscopy Control Slides
Mouse cell lines (A-20, EL4, L1210, NIH/3T3, P815 and RAW 264.7).
Application Note: Mouse Cell Array is provided for immunocytochemical analysis of protein and protein-protein interaction. Procedures for immunocytochemistry can be directly applied.
Ordering information: Slides are packaged five per pack.