Would you like to log in again?
L1210 Cell Slide, ProSci Incorporated
Supplier: Prosci
17-204
10126-914PK 108.27 USD InStock
10126-914
L1210 Cell Slide, ProSci Incorporated
Slides and Cover Glasses Microscope Slides
L1210 (lymphocytic leukemia).
Application Note: L1210 cell slide is provided for immunocytochemical analysis of protein and protein-protein interaction. Procedures for immunohistochemistry can be directly applied.
Ordering information: Slides are packaged five per pack.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty