RAW 264.7 Cell Slide, ProSci Incorporated
Supplier: ProSci
17-203
10126-912PK 158.19 USD
10126-912
RAW 264.7 Cell Slide, ProSci Incorporated
Slides Microscopy Control Slides
RAW 264.7 (Abelson murine leukemia virus-induced tumor).
Application Note: RAW 264.7 cell slide is provided for immunocytochemical analysis of protein and protein-protein interaction. Procedures for immunohistochemistry can be directly applied.
Ordering information: Slides are packaged five per pack.