A-431 Cell Slide, ProSci Incorporated
Supplier: ProSci
17-002
10126-886PK 158.19 USD
10126-886
A-431 Cell Slide, ProSci Incorporated
Slides Microscopy Control Slides
A-431 (epidermis; epidermoid carcinoma).
Application Note: A-431 cell slide is provided for immunocytochemical analysis of protein and protein-protein interaction. Procedures for immunohistochemistry can be directly applied.
Ordering information: Slides are packaged five per pack.