Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Skin Tissue Slide (Squamous Cell Carcinoma), ProSci Incorporated

Supplier: Prosci
12-704
Formalin-fixed, paraffin-embedded tissue is useful for a variety of applications including the targeting of proteins, RNA, and DNA by means of Immunohistochemistry (IHC), in-situ hybridization (ISH) and fluorescence in-situ hybridization (FISH), respectively

Application Note: ProSci’s human tissue slides can be used with any antibody shown to be effective in formalin-fixed tissue using a standard immunostaining protocol. In order to avoid false negative results from DNA or RNA sequences not being present in a 4µm cellular cross-section, 10µm tissue sections are recommended. 10µm single tissue slides are available upon request. This product has not been evaluated for effectiveness in ISH or FISH applications.

Some tissue types may require extra attention as they may contain high levels of endogenous biotin, peroxidase, or autofluorescence. Please Note: ProSci now has over 700 different lots available for a wide range of tissues. Many of these lots are not listed online. If you don't see what you are looking for please call us.

Ordering information: Slides are packaged five per pack.




 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Skin Tissue Slide (Squamous Cell Carcinoma) 12-704 10126-772 Pack of 1 Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Slide Type Human Body
Tested Applications IHC
Storage Conditions Stable for up to two years when stored in a cool, dry lab area
Species Human
Tissue/Cell Type Skin
General Diagnosis Skin Tissue Slide (Squamous Cell Carcinoma)
Fixation Formalin-fixed, paraffin-embedded

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty