Would you like to log in again?
Matched Pair Tissue Slides - Cervix Tissue, ProSci Incorporated
Supplier: Prosci
11-300-MPPT
10126-872PK 131.07 USD InStock
10126-872
Matched Pair Tissue Slides - Cervix Tissue, ProSci Incorporated
Slides and Cover Glasses Microscope Slides
The Matched Pair Paraffin Tissue (MPPT) slides are designed for identifying tumor-specific/metastasis genes or proteins
Application Note: Immunohistochemistry (IHC), immunofluorescence (IF), in-situ hybridization (ISH) and fluorescence in-situ hybridization (FISH). 10 µm tissue sections are recommended for ISH applications to avoid false negative results. 10 µm single tissue slides are available upon request. This product has not been evaluated for effectiveness in ISH or FISH applications.
Slices from normal and malignant tissues are mounted on each MPPT slide which can then be treated as a single histological slide for H&E staining, immunohistochemistry, or in-situ hybridization. This format allows a rapid analysis of protein expression and localization across normal and cancerous tissue.
Ordering information: Slides are packaged five per pack.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty