Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Jackson Safety® G60 Level 2 Polyurethane Coated Cut Resistant Gloves, Kimberly-Clark Professional®

Supplier: KIMBERLY CLARK CORP

JACKSON SAFETY®
38639 38638 38636 38635 38637
89410-120CS 167.41 USD InStock
89410-120 89410-118 89410-114 89410-112 89410-116
Jackson Safety® G60 Level 2 Polyurethane Coated Cut Resistant Gloves, Kimberly-Clark Professional®
Gloves
Add cut resistance to your workers’ gloves without compromising dexterity with our Jackson Safety* G60 Level 2 Polyurethane Coated Cut Resistant Gloves

  • CPPT 672 grams
  • 15-gauge Kevlar® liner
  • Comfortable fit
  • Excellent dexterity for improved productivity
  • Oil resistant
  • Suitable for intermittent head up tp 212°F
  • Patent pending
  • Contains natural rubber latex

CAUTION: This Product Contains Natural Rubber Latex Which May Cause Allergic Reactions.

They are a great choice for working environments that pose a risk of minor cuts and abrasions. These gloves are typically used in metal fabrication, glass handling, automotive and aerospace assembly, as well as stamping and assembly.

Certifications: ANSI/EN 388 Level 2 cut resistance. ASTM F1790 Level 2 (500 g minimum).

Ordering information: Gloves are packaged 12 pairs per bag with 1 bag per case.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty