Multiple Species Tissue Slides - Testis Tissue, ProSci Incorporated
Supplier: ProSci
11-700-MSTA
10126-796PK 185.71 USD
10126-796
Multiple Species Tissue Slides - Testis Tissue, ProSci Incorporated
Slides Microscopy Control Slides
The Multiple Species Tissue Array (MSTA) slides were designed to study protein expression patterns in different cells and tissues from multiple species
Application Note: Immunohistochemistry (IHC), immunofluorescence (IF), in situ hybridization (ISH) and fluorescence in situ hybridization (FISH). 10µm tissue sections are recommended for ISH applications to avoid false negative results. 10µm single tissue slides are available upon request. This product has not been evaluated for effectiveness in ISH or FISH applications.
Tissue slices from three different species are mounted on each MSTA slide which can then be treated as a single histological slide for H&E staining, immunohistochemistry, or in situ hybridization. This format allows a rapid analysis of protein expression and localization across different species. MSTA slides can also be used to quickly determine the species reactivity of a given antibody.
Ordering information: Slides are packaged five per pack.