Matched Pair Tissue Slides - Spleen Tissue, ProSci Incorporated
Supplier: ProSci
10-900-MPPT
10126-882PK 185.71 USD
10126-882
Matched Pair Tissue Slides - Spleen Tissue, ProSci Incorporated
Slides Microscopy Control Slides
The Matched Pair Paraffin Tissue (MPPT) slides are designed for identifying tumor-specific/metastasis genes or proteins
Application Note: Immunohistochemistry (IHC), immunofluorescence (IF), in-situ hybridization (ISH) and fluorescence in-situ hybridization (FISH). 10 µm tissue sections are recommended for ISH applications to avoid false negative results. 10 µm single tissue slides are available upon request. This product has not been evaluated for effectiveness in ISH or FISH applications.
Slices from normal and malignant tissues are mounted on each MPPT slide which can then be treated as a single histological slide for H&E staining, immunohistochemistry, or in-situ hybridization. This format allows a rapid analysis of protein expression and localization across normal and cancerous tissue.
Ordering information: Slides are packaged five per pack.