Would you like to log in again?
Breast Tissue Slide (Well Differentiated Carcinoma), ProSci Incorporated
Supplier: Prosci
10-012
10126-934PK 108.27 USD InStock
10126-934
Breast Tissue Slide (Well Differentiated Carcinoma), ProSci Incorporated
Slides and Cover Glasses Microscope Slides
Formalin-fixed, paraffin-embedded tissue is useful for a variety of applications including the targeting of proteins, RNA, and DNA by means of Immunohistochemistry (IHC), in-situ hybridization (ISH) and fluorescence in-situ hybridization (FISH), respectively
Application Note: ProSci’s human tissue slides can be used with any antibody shown to be effective in formalin-fixed tissue using a standard immunostaining protocol. In order to avoid false negative results from DNA or RNA sequences not being present in a 4 µm cellular cross-section, 10 µm tissue sections are recommended. 10 µm single tissue slides are available upon request. This product has not been evaluated for effectiveness in ISH or FISH applications.
Some tissue types may require extra attention as they may contain high levels of endogenous biotin, peroxidase, or autofluorescence. Please Note: ProSci now has over 700 different lots available for a wide range of tissues. Many of these lots are not listed online. If you don't see what you are looking for please call us.
Ordering information: Slides are packaged five per pack.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty