Would you like to log in again?
VWR Life Science Seradigm FB Essence
Supplier: VWR
3100-050H 3100-500GH 3100-500H 3100-500G 3100-050 3100-500
10799-384EA 59.04 USD InStock
10799-384 10805-182 10799-390 10805-180 10805-184 10803-034
VWR Life Science Seradigm FB Essence
Sera (Serum)
FB Essence is a nutritionally rich, cost-effective alternative to Fetal Bovine Serum (FBS) and is proven effective across a broad range of cell types and origins, including both suspension and adherent cell types and recognized finicky cell lines.
  • 100% US Origin
  • Cost effective alternative to FBS
  • Effective across a broad range of cell types
FB Essence contains FBS, Bovine Calf Serum, Equine Serum, and a proprietary blend of supplements, vitamins, minerals, and growth factors.
VWR Life Science Seradigm is a fully-integrated supplier of animal sera that provides the cell culture community with access to the most reliable supply of high performance, exceptional quality Fetal Bovine Serum (FBS) and cost-effective FBS alternatives. Our approach to sourcing and manufacturing is refreshingly unique and distinct from the competition, resulting in product performance and quality unlike any other.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty