Anti-HDAC Family Rabbit Polyclonal Antibody

Supplier: BioVision
K333-11-30
10013-682EA 1278.42 USD
10013-682
Anti-HDAC Family Rabbit Polyclonal Antibody
A convenient set of eleven HDAC family antibodies. Histone deacetylase (HDAC) family proteins play an important role in modulating the eukaryotic chromatin structure.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: HDAC Family
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR