Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SGK Rabbit Polyclonal Antibody

Supplier: BioVision

3459-100
10005-588EA 252.35 USD InStock
10005-588
Anti-SGK Rabbit Polyclonal Antibody
Antibodies
Serum and glucocorticoid-inducible kinase (SGK) is a serine/threonine kinase that is closely related to Akt. SGK is rapidly induced in response to a variety of stimuli, including serum, hormones, and osmotic shock, etc. SGK has been involved in modulating anti-apoptotic and cell cycle regulation, as well as in activating certain potassium, sodium and chloride channels.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml

Type: Primary
Antigen: SGK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty