Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Superoxide Dismutase Rabbit Polyclonal Antibody

Supplier: BioVision

3458-100
10005-584EA 262.65 USD InStock
10005-584
Anti-Superoxide Dismutase Rabbit Polyclonal Antibody
Antibodies
SOD (Superoxide Dismutase) is a well characterized cytosolic scavenger of oxygen free radicals that requires copper and zinc binding to potentiate its enzymatic activity. Enzymatically, SOD-1 facilitates the dismutation of oxygen radicals to hydrogen peroxide, and it also catalyzes prooxidant reactions, which include the peroxidase activity and hydroxyl radical generating activity. Defects in the gene encoding SOD-1 have been implicated in the progression of neurological diseases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 20 µg/ml; Immunohistochemistry: 20 µg/ml, frozen & paraffin

Type: Primary
Antigen: Superoxide Dismutase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty