Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKG Rabbit Polyclonal Antibody

Supplier: BioVision

3452-100
10003-988EA 272.95 USD InStock
10003-988
Anti-PKG Rabbit Polyclonal Antibody
Antibodies
PKGs are cyclic GMP-dependent protein kinases (also designed cGKs) that are classified into two types, PKGI and PKGII. Studies have shown that cGKs and cyclic AMP-dependent kinases (cAKs) are highly homologous protein kinase families with similar substrate specificities. Phosphorylation of cellular proteins by both families of kinases leads to alterations in calcium mobilization, protein phosphatase activity, ion channel function, gene transcription, smooth muscle contractility and platelet aggregation. However, recent studies using mice deficient in PKGs have shown that cGMP kinases regulate very specifically distinct pathways which are separate from those used by cyclic AMP kinases.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PKG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty