Anti-PKC Rabbit Polyclonal Antibody

Supplier: BioVision
3451-100
10003-986EA 379.44 USD
10003-986
Anti-PKC Rabbit Polyclonal Antibody
PKC (Protein kinase C) is a calcium-dependent and phospholipid-dependent enzyme that is activated in vivo by the lipid diacylglycerol. PKC has been shown to be involved in signal transduction and various aspects of neoplastic transformation. PKC contains an amino terminal regulatory domain and a carboxy terminal catalytic domain joined by a hinge region. Cleavage of protein kinase C in vitro results in the generation of two functional fragments corresponding to these two domains.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: PKC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR