Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKC Rabbit Polyclonal Antibody

Supplier: BioVision

3450-100
10003-982EA 272.95 USD InStock
10003-982
Anti-PKC Rabbit Polyclonal Antibody
Antibodies
PKC (Protein kinase C) is a calcium-dependent and phospholipid-dependent enzyme that is activated in vivo by the lipid diacylglycerol. PKC has been shown to be involved in signal transduction and various aspects of neoplastic transformation. PKC contains an amino terminal regulatory domain and a carboxy terminal catalytic domain joined by a hinge region. Cleavage of protein kinase C in vitro results in the generation of two functional fragments corresponding to these two domains.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PKC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty