Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PXN Rabbit Polyclonal Antibody

Supplier: BioVision

3448-100
10003-976EA 272.95 USD InStock
10003-976
Anti-PXN Rabbit Polyclonal Antibody
Antibodies
Paxillin is one of the key components of integrin signaling. Tyrosine phosphorylation of paxillin is required for integrin-mediated cytoskeletal reorganization. Paxillin is phosphorylated by another focal adhesion component, focal adhesion kinase (FAK), at tyrosine 118. Phospho-Tyr118 may provide a docking site for recruitment of other signaling molecules to focal adhesions.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: PXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty