Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAR4 Rabbit Polyclonal Antibody

Supplier: BioVision

3447-100
10003-972EA 252.35 USD InStock
10003-972
Anti-PAR4 Rabbit Polyclonal Antibody
Antibodies
Five ionomycin-inducible complementary cDNAs, designated PAR1, 2, 3, 4 and 5, have been isolated from the prostate cancer cell line AT-3. Nucleotide sequencing identified PAR1 as the rat homolog of monkeyP-1, PAR2 as the injury-inducible gene HB-EGF, and PAR3 as the serum-induced gene cyr-61. PAR4 and PAR5 sequences were not found to correspond to any previously described proteins. PAR4 (prostate apoptosis response-4) is specifically expressed by cells entering apoptosis and is not inducible by growth factor stimulation, oxidative stress and necrosis, or growth arrest. The PAR4 gene encodes a 38 kDa protein with a putative nuclear localization signal and carboxy terminal leucine zipper.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PAR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Horse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty