Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR3 Rabbit Polyclonal Antibody

Supplier: BioVision

3445R-100
10004-070EA 252.35 USD InStock
10004-070
Anti-TLR3 Rabbit Polyclonal Antibody
Antibodies
To date, at least ten members of the Toll family have been identified. TLRs recognize molecular patterns associated with microbial pathogens and induce antimicrobial immune response. TLR3 recognizes double-stranded (ds) RNA, induces the activation of NF-kB, throμgh MyD88-dependent and -independent pathways, and the production of type I interferons (IFNs). Similar to several other members of the TLR family, TLR3 has been reported to be expressed at a very low level on the surface of human fibroblast cell lines.

Recommended Dilutions: Western Blot: 0.5-4 g/ml

Type: Primary
Antigen: TLR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty