Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAK4 Rabbit Polyclonal Antibody

Supplier: BioVision

3444-100
10004-066EA 252.35 USD InStock
10004-066
Anti-PAK4 Rabbit Polyclonal Antibody
Antibodies
The PAK (p21-activated kinase) family of serine/threonine kinases plays an important role in multiple cellular processes, including cytoskeletal reorganization, MAPK signaling, apoptotic signaling, etc. Several mechanisms that induce PAK activation have been reported. Binding of Rac/cdc42 to the CRIB (or PBD) domain at the N-terminal region of PAK causes autophosphorylation and conformational change of PAK. More recently identified members PAK4, PAK5 and PAK6 have lower sequence similarity with PAK1-3 in the regulatory N-terminal region. It has been demonstrated that phosphorylation of serine 474 of PAK4 may play a pivotal role in the activity and function of PAK4 kinase

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PAK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty