Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UPAR Goat Polyclonal Antibody

Supplier: BioVision

3440-100
10004-056EA 303.85 USD InStock
10004-056
Anti-UPAR Goat Polyclonal Antibody
Antibodies
uPAR (Urokinase plasminogen activator receptor) is a 55 kDa glycoprotein I-anchored surface receptor specific for urokinase plasminogen activator (uPA). Upon binding to uPAR, uPA converts the surface bound, large serum β-globulin, plasminogen to plasmin. Plasmin, which is also designated fibrinolysin, is a trypsin-like enzyme that acts on Arg-Lys bonds and induces pericellular proteolysis in fibrin and fibrinogen, and thereby contributes to the systematic activation of the coagulation cascade. uPA and uPAR are known to be overexpressed in mesenchymal and epithelial tumor cells and are required for tumor invasion and metastasis.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml; Immunoprecipitation: 5-10 µg/ml; Neutralization: 2-4 µg/ml

Type: Primary
Antigen: UPAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty