Anti-JMJD1C Rabbit Polyclonal Antibody

Supplier: BioVision
3439-100
10003-964EA 309.39 USD
10003-964
Anti-JMJD1C Rabbit Polyclonal Antibody
JMJD1C is a member of the jumonji (jmj) domain containing gene family of histone demethylases that plays a role in chromatin regulation and influences transcriptional activation and suppression. Some recently characterized members of the jmj family include JARID1A/RBP2, JARID1C, JMJD1A, JMJD1B, JMJD1C, JMJD2A, JMJD2C and JMJD2D. JMJD1C has been identified as a nuclear-receptor coactivator and is a candidate gene for autism.

Recommended Dilutions: Western Blot: 1 - 4 µg/ml

Type: Primary
Antigen: JMJD1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR