Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK Rabbit Polyclonal Antibody

Supplier: BioVision

3438-100
10003-962EA 355.35 USD InStock
10003-962
Anti-MAPK Rabbit Polyclonal Antibody
Antibodies
p38 MAP kinase is the mammalian homologue of the yeast HOG kinase and participates in a
cascade controlling cellular responses to cytokines and stress. Like the SAPK/JNK pathway,
p38 MAP kinase is activated by a variety of cellular stresses including inflammatory cytokines,
UV light and growth factors, etc. Activated p38 MAP kinase has been shown to phosphorylate
and activate MAPKAP kinase-2 and to phosphorylate the transcription factors ATF-2 and Max.

Recommended Dilutions: Western Blot: 0.5-4 μg/ml; Immunoprecipitation: 10-20 mg/ml; Immunohistochemistry: 10-20 μg/ml

Type: Primary
Antigen: MAPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty