Anti-LIMK1 Rabbit Polyclonal Antibody

Supplier: BioVision
3433-100
10003-958EA 327.58 USD
10003-958
Anti-LIMK1 Rabbit Polyclonal Antibody
Antibodies
LIM kinases (LIMK1 and LIMK2) are serine/threnine kinases that have two zinc finger motifs, known as LIM motifs in their N-terminal regulatory domain. LIM kinases are involved in actin cytoskeleton regulation throμgh Rho-family GTPases and downstream kinases PAKs and ROCK. PAK1 and ROCK phosphorylate LIMK1 and LIMK2, which increases the activity of the kinases. Activated LIM kinases inhibit the actin depolymerization activity of cofilin by phosphorylation at the N-terminus of Cofilin.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: LIMK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR