Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MLK3 Rabbit Polyclonal Antibody

Supplier: BioVision

3430-100
10003-956EA 272.95 USD InStock
10003-956
Anti-MLK3 Rabbit Polyclonal Antibody
Antibodies
MLK3 (Mixed lineage kinase 3) is a serine/threonine kinase that has an amino-terminal SH3 domain followed by the kinase domain and two leucine zippers, a cdc42/Rac1 binding (CRIB) domain and several other domains/motifs at the carboxy-terminal region. MLK3 functions as a MAPKKK of the SAPK/JNK stress pathway by directly phosphorylating SEK1/MKK4 and MKK7, althoμgh it is controversial whether MLK3 is involved in p38 stress pathway activation. MLK3 also functions as an IkB kinase and mediates the activation of the transcriptional factor NF-kB stimulated by CD3/CD28, sμggesting a role for MLK3 in immune and inflammatory responses.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty