Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MSH2 Rabbit Polyclonal Antibody

Supplier: BioVision

3429-100
10003-954EA 252.35 USD InStock
10003-954
Anti-MSH2 Rabbit Polyclonal Antibody
Antibodies
DNA mismatch repair genes have been found to be associated with hereditary nonpolyposis colorectal cancer (HNPCC). Inherited mutations in the MSH2 were demonstrated at high frequency in HNPCC and were shown to be associated with microsatellite instability. The demonstration that 10 to 45% of pancreatic, gastric, breast, ovarian and small cell lung cancers also display microsatellite instability sμggests that DNA mismatch repair is not restricted to HNPCC tumors but is a common feature in tumor initiation or progression.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunoprecipitation: 15-25 µg/ml; and Immunohistochemistry: 15-25 µg/ml

Type: Primary
Antigen: MSH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty