Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Neurexin Rabbit Polyclonal Antibody

Supplier: BioVision

3428-100
10003-950EA 272.95 USD InStock
10003-950
Anti-Neurexin Rabbit Polyclonal Antibody
Antibodies
Neurexins comprise a family of neuronal cell surface proteins present primarily in the brain. Neurexins are thoμght to serve as signaling receptors and may play a role in axon guidance and synaptogenesis. The cytoplasmic tail of neurexin interacts with the synaptic vesicle protein synaptotagmin and binds to CASK, a protein with homology to the PSD95 family of synaptic junction proteins.

Recommended Dilutions: Western Blot: 1:200-1000

Type: Primary
Antigen: Neurexin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty