Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ENOS Rabbit Polyclonal Antibody

Supplier: BioVision

3426R-100
10003-948EA 252.35 USD InStock
10003-948
Anti-ENOS Rabbit Polyclonal Antibody
Antibodies
eNOS (Endothelial nitric-oxide synthase) is an important enzyme in the cardiovascular system. eNOS is regulated by phosphorylation at multiple sites. The two most thoroμghly studied sites are the activation site Ser1177 and the inhibitory site Thr495. Several protein kinases including Akt/PKB, PKA and AMPK activate eNOS by phosphorylating Ser1177 in response to various stimuli. In contrast, bradykinin and hydrogen peroxide activate eNOS activity by promoting Thr495 dephosphorylation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ENOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty