Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRS1 Rabbit Polyclonal Antibody

Supplier: BioVision

3424-100
10003-940EA 252.35 USD InStock
10003-940
Anti-IRS1 Rabbit Polyclonal Antibody
Antibodies
IRS-1 (Insulin receptor substrate-1) is phosphorylated in response to stimulation of cells by insulin. Upon ligand-induced phosphorylation, IRS-1 immediately associates with a series of SH2 domain-containing signaling intermediates such as PI3-kinase, GRB2, Syp and Nck. The level of diversity in structure and function of the SH2-containing proteins which binds activated IRS-1 is important for responsiveness to insulin. IRS phosphorylation elicits the growth and metablic effects of insulin growth factors and insulin by allowing glucose uptake, glycogen synthesis and mitogenic activity. IRS-1 migrates between 170 and 185 kDa and is expressed in most cell types.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty