Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ILK1 Rabbit Polyclonal Antibody

Supplier: BioVision

3423-100
10003-936EA 252.35 USD InStock
10003-936
Anti-ILK1 Rabbit Polyclonal Antibody
Antibodies
ILKs (Integrin-linked kinases) in combination with integrins and growth factors regulate cell survival, cell cycle, cell-cell adhesion and cell motility. ILK functions as a scaffold bridging the extra-cellular matrix (ECM) and growth factor receptors to the actin cytoskeleton throμgh interactions with integrin, PINCH (which links ILK to the RTKs via Nck2), CH-ILKBP and affixin. ILK phosphorylates several cellular targets including Akt, GSK-3, myosin light chain 2, as well as affixin. These phosphorylation events are key regulatory steps in modulating the activities of the targets.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml, frozen & paraffin

Type: Primary
Antigen: ILK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty