Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ICAM1 Rabbit Polyclonal Antibody

Supplier: BioVision

3422R-100
10003-932EA 288.4 USD InStock
10003-932
Anti-ICAM1 Rabbit Polyclonal Antibody
Antibodies
Cell adhesion molecules (CAMs) are a family of closely related cell surface glycoproteins involved in cell-cell interactions during growth and are thoμght to play an important role in embryogenesis and development. The family contains NCAM (for neuronal cell adhesion molecule), ICAM (for intercellular adhesion molecule), VCAM (vascular cell adhesion molecule), PECAM-1 (platelet/endothelial cell adhesion molecule-1), etc. ICAM-1, also referred to as CD54, is an integral membrane protein of the immunoglobulin superfamily and recognizes the B2a1 and B2aM integrins.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty