Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GADD153 Rabbit Polyclonal Antibody

Supplier: BioVision

3421-100
10003-928EA 272.95 USD InStock
10003-928
Anti-GADD153 Rabbit Polyclonal Antibody
Antibodies
GADD153 has been described as a growth arrest and DNA damage-inducible gene that encodes a C/EBP-related nuclear protein. Expression of GADD153 is induced by a variety of cellular stresses, including nutrient deprivation and metabolic perturbations. GADD153 functions to block cells in G1 to S phase in cell cycle progression and acts by dimerizing with other C/EBP proteins to direct GADD153 dimers away from “classical” C/EBP binding sites. Thus GADD153 acts as a negative modulator of C/EBP-like proteins in certain terminally differentiated cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: GADD153
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty