Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTSW Mouse Monoclonal Antibody [clone: BV39-2B]

Supplier: BioVision

3412-100
10004-042EA 303.85 USD InStock
10004-042
Anti-CTSW Mouse Monoclonal Antibody [clone: BV39-2B]
Antibodies
The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in various cellular events such as peptide biosynthesis and protein degradation.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunohistochemistry: 20-30 µg/ml, tested on frozen section

Type: Primary
Antigen: CTSW
Clonality: Monoclonal
Clone: BV39-2B
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty