Anti-CTSV Mouse Monoclonal Antibody [clone: BV55-1]

Supplier: BioVision
3411-100
10004-040EA 361.93 USD
10004-040
Anti-CTSV Mouse Monoclonal Antibody [clone: BV55-1]
The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in various cellular events such as peptide biosynthesis and protein degradation.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunohistochemistry: 20-30 µg/ml, paraffin section after microwave treatment

Type: Primary
Antigen: CTSV
Clonality: Monoclonal
Clone: BV55-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR